Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017915.2 | NP_001017915.1 | 1 | 571 | ||
NM_005541.4 | NP_005532.2 | 1 | 571 | |||
XM_017004004.1 | XP_016859493.1 | 1 | 194 | |||
GenBank mRNA | AK298651.1 | - | 1 | 200 | ||
AK309644.1 | - | 1 | 196 | |||
BC099920.1 | - | 1 | 182 | 6304992 | ||
BC113580.1 | - | 1 | 119 | 8322632 | ||
BC113582.1 | - | 1 | 119 | 8322634 | ||
BE247227.1 | - | 1 | 216 | |||
BM800110.1 | - | 208 | ||||
U50041.1 | - | 1 | 180 | |||
U53470.1 | - | 1 | 189 | |||
U57650.1 | - | 1 | 571 | |||
U84400.1 | - | 1 | 140 | |||
X98429.1 | - | 1 | 199 |