Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016058.2 | NP_057142.1 | 4 | 509 | ||
XM_005264343.3 | XP_005264400.1 | 5 | 1285 | |||
XM_006712026.3 | XP_006712089.1 | 6 | 1100 | |||
XM_006712027.3 | XP_006712090.1 | 5 | 1214 | |||
XM_011532877.2 | XP_011531179.1 | 5 | 1211 | |||
XM_011532878.2 | XP_011531180.1 | 5 | 558 | |||
XM_011532879.2 | XP_011531181.1 | 5 | 872 | |||
XM_011532880.2 | XP_011531182.1 | 5 | 869 | |||
XM_017004232.1 | XP_016859721.1 | 6 | 1098 | |||
XM_017004233.1 | XP_016859722.1 | 5 | 1295 | |||
XM_017004234.1 | XP_016859723.1 | 4 | 1181 | |||
XM_017004235.1 | XP_016859724.1 | 5 | 963 | |||
GenBank mRNA | AF060921.1 | - | 4 | 448 | ||
AF151879.1 | - | 4 | 504 | |||
AK097254.1 | - | 3 | 2061 | |||
AK291071.1 | - | 4 | 440 | |||
AK295239.1 | - | 4 | 386 | |||
AY157986.1 | - | 4 | 515 | |||
AY157987.1 | - | 3 | 299 | |||
AY643713.1 | - | 4 | 460 | |||
BC029492.1 | - | 4 | 461 | 5271898 | ||
DA688967.1 | - | 4 | 509 |