Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139314.2 | NP_647475.1 | 4 | 793 | ||
XM_005272484.3 | XP_005272541.1 | 4 | 791 | |||
GenBank mRNA | AB056477.1 | - | 4 | 767 | ||
AF153606.1 | - | 4 | 748 | |||
AF169312.1 | - | 4 | 662 | |||
AF202636.1 | - | 4 | 770 | |||
AK222489.1 | - | 4 | 765 | |||
AK222524.1 | - | 4 | 763 | |||
AK303269.1 | - | 5 | 694 | |||
AY358275.1 | - | 4 | 761 | |||
BC023647.2 | - | 4 | 737 | 5088323 |