Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287054.1 | NP_001273983.1 | 8 | 1101 | ||
NM_001287055.1 | NP_001273984.1 | 4 | 404 | |||
NM_003439.2 | NP_003430.1 | 6 | 1072 | |||
XM_011516559.2 | XP_011514861.1 | 6 | 1627 | |||
GenBank mRNA | AF454940.1 | - | 6 | 1043 | ||
AK054676.1 | - | 8 | 1074 | |||
AK074116.1 | - | 6 | 1030 | |||
AK091471.1 | - | 4 | 373 | |||
AK123012.1 | - | 1 | 1074 | |||
AL833578.1 | - | 6 | 1048 | |||
BC022378.1 | - | 6 | 1045 | 4685873 | ||
BC112224.1 | - | 5 | 875 | 8327692 | ||
BC113697.1 | - | 5 | 875 | 8322749 | ||
BX640646.1 | - | 6 | 1045 | |||
U09848.1 | - | 2 | 115 |