Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 5, 6 | 588 | |||
GenBank mRNA | AK291385.1 | - | 5, 6 | 536 |
Gene Symbol: MIA-RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037775.1 | 7, 8 | 793 | |||
GenBank mRNA | AF087861.1 | - | 6, 7 | 574 |
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016154.4 | NP_057238.3 | 5, 6 | 588 | ||
GenBank mRNA | AF165522.1 | - | 5, 6 | 554 | ||
AF217985.1 | - | 4, 5 | 1392 | |||
AF498935.1 | - | 5, 6 | 431 | |||
BC046927.1 | - | 5, 6 | 541 | 5805921 | ||
BG706803.1 | - | 5, 6 | 596 |