Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001777.3 | NP_001768.1 | 2 | 519 | ||
NM_198793.2 | NP_942088.1 | 2 | 519 | |||
XM_005247908.1 | XP_005247965.1 | 2 | 489 | |||
XM_005247909.1 | XP_005247966.1 | 2 | 489 | |||
GenBank mRNA | AK289813.1 | - | 2 | 460 | ||
BC010016.2 | - | 2 | 421 | 4303534 | ||
BC012884.1 | - | 2 | 442 | 3857911 | ||
BC037306.1 | - | 2 | 456 | 5260986 | ||
BT006907.1 | - | 2 | 339 | |||
LN680437.1 | - | 2 | 339 | |||
X69398.1 | - | 2 | 445 | |||
Z25521.1 | - | 2 | 478 |