Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320494.1 | NP_001307423.1 | 4 | 9190 | ||
NM_016206.3 | NP_057290.2 | 4 | 9349 | |||
XM_006713138.3 | XP_006713201.1 | 4 | 9516 | |||
GenBank mRNA | AF099505.1 | - | 1 | 485 | ||
AK095826.1 | - | 1 | 1731 | |||
BC062478.1 | - | 1 | 399 |