Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177973.1 | NP_001171444.1 | 13 | 1924 | ||
NM_001177974.1 | NP_001171445.1 | 12 | 1804 | |||
NM_001177975.1 | NP_001171446.1 | 12 | 1804 | |||
NM_001177976.1 | NP_001171447.1 | 12 | 1804 | |||
NM_008363.2 | NP_032389.2 | 12 | 1804 | |||
GenBank mRNA | AF103876.1 | - | 12 | 1804 | ||
AK045958.1 | - | 12 | 1713 | |||
AK052933.1 | - | 10 | 1674 | |||
AK143694.1 | - | 12 | 1715 | |||
AK144879.1 | - | 12 | 1713 | |||
AK203259.1 | - | 2 | 231 | |||
AY184362.1 | - | 12 | 1804 | |||
AY184363.1 | - | 12 | 1804 | |||
AY184364.1 | - | 12 | 1804 | |||
BC004778.1 | - | 13 | 1916 | 3257579 | ||
U56773.2 | - | 12 | 1628 |