Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016246.2 | NP_057330.2 | 3, 4 | 469 | ||
XM_005258969.3 | XP_005259026.1 | 3, 4 | 478 | |||
XM_005258970.3 | XP_005259027.1 | 3, 4 | 480 | |||
GenBank mRNA | AF126781.1 | - | 3, 4 | 315 | ||
AK307437.1 | - | 3, 4 | 278 | |||
AY358430.1 | - | 3, 4 | 308 | |||
BC006283.2 | - | 3, 4 | 262 | 4026397 | ||
BC006294.1 | - | 3, 4 | 270 | 4025809 | ||
BI602824.1 | - | 3, 4 | 476 |