Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 29 | 3574 | ||
NM_001291721.1 | NP_001278650.1 | 28 | 3496 | |||
NM_001291722.1 | NP_001278651.1 | 30 | 3649 | |||
NM_014376.3 | NP_055191.2 | 29 | 3700 | |||
XM_011534516.2 | XP_011532818.1 | 29 | 3840 | |||
XM_017009341.1 | XP_016864830.1 | 29 | 3689 | |||
XM_017009342.1 | XP_016864831.1 | 30 | 3611 | |||
GenBank mRNA | AB032994.2 | - | 30 | 3589 | ||
AF160973.1 | - | 29 | 3483 | |||
AK223604.1 | - | 29 | 3527 | |||
AK294220.1 | - | 24 | 2943 | |||
AK295865.1 | - | 26 | 3123 | |||
AK297401.1 | - | 27 | 3306 | |||
AK303702.1 | - | 28 | 3449 | |||
AL136549.1 | - | 29 | 3573 | |||
AL161999.1 | - | 21 | 2534 | |||
BC011762.1 | - | 29 | 3700 | 3619680 | ||
BC021008.1 | - | 29 | 3700 |