Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004041.4 | NP_004032.2 | 7, 8 | 711 | ||
NM_020251.3 | NP_064647.1 | 7, 8 | 711 | |||
XM_011545034.2 | XP_011543336.1 | 8, 9 | 595 | |||
XM_011545035.2 | XP_011543337.1 | 8, 9 | 595 | |||
XM_017017750.1 | XP_016873239.1 | 8, 9 | 595 | |||
XM_017017751.1 | XP_016873240.1 | 8, 9 | 595 | |||
XM_017017752.1 | XP_016873241.1 | 7, 8 | 551 | |||
XM_017017753.1 | XP_016873242.1 | 7, 8 | 615 | |||
XM_017017754.1 | XP_016873243.1 | 7, 8 | 551 | |||
GenBank mRNA | AF084040.1 | - | 7, 8 | 487 | ||
AF084940.1 | - | 7, 8 | 487 | |||
AK289718.1 | - | 7, 8 | 541 | |||
BC003636.2 | - | 7, 8 | 564 | 3604829 | ||
BQ883950.1 | - | 7, 8 | 734 | |||
FJ348262.1 | - | 7, 8 | 487 |