Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037872.2 | NP_001032961.1 | 3 | 333 | ||
NM_001321454.1 | NP_001308383.1 | 3 | 333 | |||
NM_001321455.1 | NP_001308384.1 | 3 | 333 | |||
NM_001321458.1 | NP_001308387.1 | 3 | 333 | |||
NM_001321459.1 | NP_001308388.1 | 3 | 333 | |||
NM_001321460.1 | NP_001308389.1 | 3 | 333 | |||
NM_016316.3 | NP_057400.1 | 3 | 333 | |||
NR_135649.1 | 3 | 333 | ||||
NR_135650.1 | 3 | 333 | ||||
NR_135651.1 | 3 | 333 | ||||
NR_135652.1 | 3 | 333 | ||||
NR_135653.1 | 3 | 333 | ||||
XM_017004309.1 | XP_016859798.1 | 3 | 746 | |||
XM_017004310.1 | XP_016859799.1 | 3 | 112 | |||
XM_017004311.1 | XP_016859800.1 | 3 | 112 | |||
XM_017004312.1 | XP_016859801.1 | 3 | 328 | |||
XM_017004313.1 | XP_016859802.1 | 3 | 2062 | |||
GenBank mRNA | AB047646.1 | - | 3 | 258 | ||
AF151538.1 | - | 3 | 286 | |||
AF206019.1 | - | 3 | 295 | |||
AF357886.1 | - | 3 | 251 | |||
AK002087.1 | - | 1 | 195 | |||
AK307719.1 | - | 3 | 244 | |||
BC037734.1 | - | 3 | 314 | 4824689 | ||
BC130411.1 | - | 2 | 104 | 40146442 | ||
DR000409.1 | - | 3 | 224 | |||
HY010913.1 | - | 3 | 334 |