Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004319.2 | NP_001004319.1 | 1 | 951 | ||
GenBank mRNA | AY494836.1 | - | 1 | 613 | ||
BC130596.1 | - | 1 | 578 | 40146627 | ||
BC130598.1 | - | 1 | 578 | 40146629 |