Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EWSR1 SIRNA Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
EWS, EWS-FLI1, bK984G1.4
Chromosome Location:
Chr.22: 29268009 - 29300525 on Build GRCh38
UniGene ID:
Hs.374477
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241464
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EWSR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163285.1
NP_001156757.1
1, 2
347
NM_001163286.1
NP_001156758.1
1, 2
347
NM_001163287.1
NP_001156759.1
1, 2
347
NM_005243.3
NP_005234.1
1, 2
347
NM_013986.3
NP_053733.2
1, 2
347
XM_005261389.3
XP_005261446.1
1, 2
102
XM_005261390.4
XP_005261447.1
1, 2
102
XM_011529995.2
XP_011528297.1
1, 2
74
XM_011529996.2
XP_011528298.1
1, 2
74
XM_011529997.1
XP_011528299.1
1, 2
74
XM_011529998.1
XP_011528300.1
1, 2
74
XM_011529999.2
XP_011528301.1
1, 2
74
XM_011530000.1
XP_011528302.1
1, 2
74
XM_011530001.2
XP_011528303.1
1, 2
74
XM_011530002.2
XP_011528304.1
1, 2
74
XM_017028644.1
XP_016884133.1
1, 2
102
XM_017028645.1
XP_016884134.1
1, 2
102
XM_017028646.1
XP_016884135.1
1, 2
74
XM_017028647.1
XP_016884136.1
1, 2
102
XM_017028648.1
XP_016884137.1
1, 2
102
XM_017028649.1
XP_016884138.1
1, 2
74
XM_017028650.1
XP_016884139.1
1, 2
74
XM_017028651.1
XP_016884140.1
1, 2
102
XM_017028652.1
XP_016884141.1
1, 2
74
XM_017028653.1
XP_016884142.1
1, 2
102
XM_017028654.1
XP_016884143.1
1, 2
74
XM_017028655.1
XP_016884144.1
1, 2
102
XM_017028656.1
XP_016884145.1
1, 2
102
XM_017028657.1
XP_016884146.1
1, 2
63
XM_017028658.1
XP_016884147.1
1, 2
74
XM_017028659.1
XP_016884148.1
1, 2
102
XM_017028660.1
XP_016884149.1
1, 2
102
XM_017028661.1
XP_016884150.1
1, 2
63
XM_017028662.1
XP_016884151.1
1, 2
102
XM_017028663.1
XP_016884152.1
1, 2
102
XM_017028664.1
XP_016884153.1
1, 2
57
XM_017028665.1
XP_016884154.1
1, 2
57
XM_017028666.1
XP_016884155.1
1, 2
63
GenBank mRNA
AK056309.1
-
1, 2
40
AK056681.1
-
1, 2
30
AK127624.1
-
56
AK225096.1
-
1, 2
29
AK225102.1
-
1, 2
41
BC000527.2
-
1, 2
59
2961279
BC004817.1
-
1, 2
38
3449145
BC011048.1
-
1, 2
46
4340116
BC072442.1
-
25
6047805
CR456490.1
-
1, 2
52
CX786322.1
-
1, 2
77
DA013931.1
-
1, 2
347
DA051447.1
-
1, 2
45
DB051511.1
-
1, 2
328
DB443651.1
-
1, 2
338
JX977847.1
-
1, 2
347
X66899.1
-
1, 2
62
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001283062, NM_001283061, NM_001283063, NM_007968, AK147103, and others.
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0