Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251987.1 | NP_001238916.1 | 3 | 594 | ||
NM_001256020.1 | NP_001242949.1 | 3 | 594 | |||
NM_001256021.1 | NP_001242950.1 | 3 | 594 | |||
NM_001256022.1 | NP_001242951.1 | 3 | 594 | |||
NM_006073.3 | NP_006064.2 | 3 | 594 | |||
GenBank mRNA | AJ489257.2 | - | 3 | 489 | ||
AK291867.1 | - | 3 | 451 | |||
BC029393.1 | - | 3 | 451 | |||
BC057222.1 | - | 3 | 455 | |||
BC070290.1 | - | 3 | 473 | |||
BC139910.1 | - | 3 | 410 | |||
JN900469.1 | - | 3 | 408 | |||
U18985.1 | - | 3 | 298 |