Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LETM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012318.2 | NP_036450.1 | 4 | 1017 | ||
XM_006713884.1 | XP_006713947.1 | 4 | 725 | |||
XM_017008187.1 | XP_016863676.1 | 3 | 1138 | |||
GenBank mRNA | AF061025.1 | - | 4 | 1017 | ||
AK293572.1 | - | 4 | 725 | |||
AK310563.1 | - | 2 | 171 | |||
AL133650.1 | - | 3 | 783 | |||
BC014500.1 | - | 4 | 917 | 4860194 | ||
BC021208.1 | - | 4 | 942 | 4126510 |