Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 38 | 7744 | ||
XM_005267502.2 | XP_005267559.2 | 38 | 7906 | |||
XM_011536619.2 | XP_011534921.1 | 38 | 7336 | |||
XM_011536621.2 | XP_011534923.1 | 37 | 6505 | |||
XM_011536622.1 | XP_011534924.1 | 29 | 5669 | |||
XM_017021148.1 | XP_016876637.1 | 37 | 7095 | |||
XM_017021149.1 | XP_016876638.1 | 37 | 7089 | |||
XM_017021150.1 | XP_016876639.1 | 36 | 7053 | |||
XM_017021151.1 | XP_016876640.1 | 37 | 7101 | |||
XM_017021152.1 | XP_016876641.1 | 37 | 7095 | |||
XM_017021153.1 | XP_016876642.1 | 37 | 6499 | |||
GenBank mRNA | AB032957.2 | - | 30 | 5890 | ||
AK024847.1 | - | 16 | 3573 | |||
AK125788.1 | - | 12 | 2851 | |||
AL110222.1 | - | 13 | 2262 | |||
AY254380.1 | - | 38 | 7544 | |||
AY739714.1 | - | 14 | 3058 | |||
BC011658.2 | - | 3 | 945 | |||
BC063686.1 | - | 1 | 427 |