Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPVD1 SIRNA Assays ›
Gene Symbol:
GAPVD1
Gene Name:
GTPase activating protein and VPS9 domains 1
Gene Aliases:
GAPEX5, GAPex-5, RAP6
Chromosome Location:
Chr.9: 125261794 - 125365011 on Build GRCh38
UniGene ID:
Hs.495134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241619
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAPVD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282679.1
NP_001269608.1
14
2602
NM_001282680.1
NP_001269609.1
14
2602
NM_001282681.1
NP_001269610.1
11
2207
NM_015635.3
NP_056450.2
12
2436
XM_005251901.3
XP_005251958.1
13
2466
XM_005251904.3
XP_005251961.1
13
2463
XM_006717044.3
XP_006717107.1
13
2388
XM_011518499.2
XP_011516801.1
13
2480
XM_011518500.2
XP_011516802.1
14
2597
XM_011518502.2
XP_011516804.1
13
2408
XM_011518506.2
XP_011516808.1
13
2405
XM_011518507.2
XP_011516809.1
13
2485
XM_017014600.1
XP_016870089.1
12
2290
XM_017014601.1
XP_016870090.1
12
2393
XM_017014602.1
XP_016870091.1
14
2613
XM_017014603.1
XP_016870092.1
14
2560
XM_017014604.1
XP_016870093.1
15
2662
XM_017014605.1
XP_016870094.1
12
2393
XM_017014606.1
XP_016870095.1
14
2507
XM_017014607.1
XP_016870096.1
14
2516
XM_017014608.1
XP_016870097.1
12
2326
XM_017014609.1
XP_016870098.1
14
2575
XM_017014610.1
XP_016870099.1
12
2393
XM_017014611.1
XP_016870100.1
13
2386
GenBank mRNA
AB040954.3
-
14
2528
AK000126.1
-
10
1826
AK023841.1
-
5
551
AK027049.1
-
10
1288
AK056493.1
-
12
2398
AK294542.1
-
12
2436
AK296381.1
-
13
2526
AL080196.1
-
2
218
BC010027.2
-
11
2275
BC021119.1
-
10
1926
BC114937.1
-
14
2485
40083103
BC114962.1
-
12
2321
40083104
DQ233254.1
-
11
2175
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
signal transduction
positive regulation of GTPase activity
regulation of protein transport
cell-cell adhesion
guanyl-nucleotide exchange factor activity
GTPase activator activity
GTPase activating protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0