Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077628.2 | NP_001071096.1 | 2 | 571 | ||
NM_016022.3 | NP_057106.2 | 2 | 571 | |||
NR_045035.1 | 2 | 571 | ||||
GenBank mRNA | AF151835.1 | - | 2 | 273 | ||
AF508787.1 | - | 2 | 229 | |||
AK027879.1 | - | 2 | 341 | |||
AK075295.1 | - | 2 | 290 | |||
AK075452.1 | - | 2 | 379 | |||
AK298876.1 | - | 2 | 350 | |||
AK314490.1 | - | 2 | 341 | |||
AX775955.1 | - | 2 | 307 | |||
AX775957.1 | - | 2 | 361 | |||
AY113698.1 | - | 2 | 229 | |||
AY113699.1 | - | 2 | 229 | |||
AY358951.1 | - | 2 | 375 | |||
BC001230.2 | - | 2 | 341 | 3457593 | ||
BC008732.2 | - | 2 | 328 | 3162330 | ||
BC009501.2 | - | 2 | 362 | 4040230 | ||
BC015568.2 | - | 2 | 330 | 3356401 | ||
BC017699.1 | - | 2 | 307 | 4177580 | ||
BC020590.1 | - | 2 | 361 | 4420942 |