Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016245.4 | NP_057329.3 | 2 | 463 | ||
XM_011532021.1 | XP_011530323.1 | 2 | 412 | |||
GenBank mRNA | AF126780.1 | - | 2 | 418 | ||
AF273056.1 | - | 2 | 316 | |||
AK075348.1 | - | 2 | 347 | |||
AY358553.1 | - | 2 | 330 | |||
BC008650.1 | - | 2 | 353 | 3854750 | ||
BC014327.1 | - | 2 | 329 | 3961721 | ||
BC016367.1 | - | 2 | 360 | 4133318 | ||
BC021673.1 | - | 2 | 334 | 3827935 | ||
BC036001.1 | - | 2 | 354 | 4715042 | ||
BU838518.1 | - | 545 | ||||
DA265697.1 | - | 2 | 462 |