Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 11 | 1416 | ||
NM_016836.3 | NP_058520.1 | 11 | 1425 | |||
XM_005246737.1 | XP_005246794.1 | 12 | 1184 | |||
XM_005246738.2 | XP_005246795.1 | 12 | 1519 | |||
XM_005246739.2 | XP_005246796.1 | 12 | 1389 | |||
XM_005246740.1 | XP_005246797.1 | 12 | 1175 | |||
XM_005246741.3 | XP_005246798.1 | 12 | 1386 | |||
XM_017004626.1 | XP_016860115.1 | 11 | 1462 | |||
GenBank mRNA | AK127157.1 | - | 2 | 1817 | ||
AK225887.1 | - | 12 | 1084 | |||
AK297812.1 | - | 12 | 1175 | |||
BC012992.2 | - | 11 | 1111 | 3530897 | ||
BC012993.2 | - | 11 | 1111 | 3530898 | ||
BC018951.2 | - | 11 | 1412 | 4137722 | ||
BC051889.1 | - | 11 | 998 | |||
BX648246.1 | - | 4 | 8582 | |||
D28482.1 | - | 11 | 982 | |||
KU178305.1 | - | 11 | 972 | |||
KU178306.1 | - | 12 | 1026 | |||
L11289.1 | - | 11 | 1049 | |||
X64652.1 | - | 10 | 932 | |||
X77494.1 | - | 13 | 1161 |