Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017631.5 | NP_060101.3 | 36 | 5206 | ||
XM_011532103.2 | XP_011530405.1 | 36 | 5206 | |||
GenBank mRNA | AK000042.1 | - | 8 | 1007 | ||
AK026333.1 | - | 4 | 472 | |||
AK223184.1 | - | 8 | 1007 | |||
AK225191.1 | - | 8 | 994 | |||
AL137464.1 | - | 3 | 392 | |||
BC020601.1 | - | 4 | 422 | |||
BC038115.2 | - | 36 | 5151 | 5261001 | ||
BC074781.2 | - | 8 | 985 | |||
BX648655.1 | - | 16 | 2048 |