Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039395.3 | NP_001034484.3 | 3 | 355 | ||
XM_006716795.2 | XP_006716858.1 | 2 | 208 | |||
XM_006716798.3 | XP_006716861.1 | 3 | 213 | |||
XM_006716799.3 | XP_006716862.1 | 2 | 315 | |||
XM_006716800.2 | XP_006716863.1 | 2 | 163 | |||
XM_006716801.2 | XP_006716864.1 | 3 | 355 | |||
XM_011517952.1 | XP_011516254.1 | 2 | 151 | |||
XM_011517954.1 | XP_011516256.1 | 2 | 295 | |||
XM_011517956.1 | XP_011516258.1 | 2 | 208 | |||
XM_011517957.1 | XP_011516259.1 | 2 | 295 | |||
XM_011517958.1 | XP_011516260.1 | 1 | 69 | |||
XM_017014881.1 | XP_016870370.1 | 2 | 208 | |||
XM_017014882.1 | XP_016870371.1 | 2 | 208 | |||
XM_017014883.1 | XP_016870372.1 | 2 | 208 | |||
XM_017014884.1 | XP_016870373.1 | 2 | 163 | |||
XM_017014885.1 | XP_016870374.1 | 2 | 208 | |||
XM_017014886.1 | XP_016870375.1 | 2 | 111 | |||
XM_017014887.1 | XP_016870376.1 | 2 | 193 | |||
GenBank mRNA | AK000920.1 | - | 3 | 189 | ||
AK295831.1 | - | 3 | 347 | |||
AK301790.1 | - | 2 | 308 | |||
BC034293.1 | - | 2 | 282 | 5167647 | ||
BC036349.1 | - | 3 | 207 | |||
BX649030.1 | - | 2 | 258 | |||
DA234263.1 | - | 3 | 355 |