Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 20 | 1972 | ||
XM_011543530.1 | XP_011541832.1 | 20 | 2149 | |||
XM_011543531.2 | XP_011541833.1 | 20 | 2149 | |||
XM_011543532.1 | XP_011541834.1 | 20 | 2149 | |||
XM_011543533.1 | XP_011541835.1 | 19 | 2065 | |||
XM_017009689.1 | XP_016865178.1 | 19 | 2065 | |||
GenBank mRNA | AB030240.1 | - | 20 | 1704 | ||
AK096057.1 | - | 9 | 654 | |||
BC114632.1 | - | 20 | 1798 | |||
CB048933.1 | - | 7 | 527 |