Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144903.1 | NP_001138375.1 | 9 | 1224 | ||
NM_023003.3 | NP_075379.2 | 10 | 1317 | |||
XM_011521680.2 | XP_011519982.1 | 11 | 1366 | |||
XM_017022323.1 | XP_016877812.1 | 10 | 1369 | |||
XM_017022324.1 | XP_016877813.1 | 9 | 1253 | |||
XM_017022326.1 | XP_016877815.1 | 10 | 1231 | |||
XM_017022327.1 | XP_016877816.1 | 10 | 1423 | |||
XM_017022328.1 | XP_016877817.1 | 7 | 992 | |||
GenBank mRNA | AF255922.1 | - | 10 | 1265 | ||
AK223246.1 | - | 10 | 1262 | |||
AK292100.1 | - | 10 | 1242 | |||
AK295568.1 | - | 10 | 1206 | |||
BC032007.1 | - | 6 | 828 | |||
BC063487.1 | - | 7 | 997 | 5240735 |