Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPAG16 SIRNA Assays ›
Gene Symbol:
SPAG16
Gene Name:
sperm associated antigen 16
Gene Aliases:
PF20, WDR29
Chromosome Location:
Chr.2: 213284369 - 214410501 on Build GRCh38
UniGene ID:
Hs.743365
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241833
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPAG16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025436.2
NP_001020607.1
4
481
NM_024532.4
NP_078808.3
4
481
NR_047659.1
6
656
XM_006712746.2
XP_006712809.1
4
432
XM_011511814.1
XP_011510116.1
3
529
XM_011511815.1
XP_011510117.1
4
432
XM_011511816.2
XP_011510118.1
4
432
XM_011511817.1
XP_011510119.1
4
432
XM_011511818.1
XP_011510120.1
4
432
XM_011511819.2
XP_011510121.1
3
1180
XM_011511820.1
XP_011510122.1
4
432
XM_011511824.1
XP_011510126.1
4
432
XM_011511827.1
XP_011510129.1
4
432
XM_011511828.1
XP_011510130.1
4
432
XM_011511829.1
XP_011510131.1
4
432
XM_011511831.1
XP_011510133.1
4
432
XM_011511832.1
XP_011510134.1
4
432
XM_011511835.2
XP_011510137.1
3
524
XM_011511837.2
XP_011510139.1
4
432
XM_017004896.1
XP_016860385.1
5
561
XM_017004897.1
XP_016860386.1
2
3178
XM_017004899.1
XP_016860388.1
4
432
XM_017004900.1
XP_016860389.1
4
423
XM_017004901.1
XP_016860390.1
4
413
GenBank mRNA
AF310672.1
-
4
418
AF426740.1
-
4
418
AF490390.1
-
4
411
AK026377.1
-
4
441
AK095036.1
-
3
529
AK310883.1
-
4
380
AL832962.1
-
4
370
BC009614.1
-
4
442
BC025379.1
-
6
661
4820489
BC067756.1
-
4
468
BC100282.1
-
4
429
CR457359.1
-
4
376
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sperm axoneme assembly
axoneme assembly
cilium assembly
microtubule sliding
cilium movement involved in cell motility
cell motility in response to calcium ion
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0