Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAP130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145928.1 | NP_001139400.1 | 12 | 1678 | ||
NM_024545.3 | NP_078821.2 | 12 | 1678 | |||
XM_005263767.3 | XP_005263824.1 | 12 | 1649 | |||
XM_005263768.3 | XP_005263825.1 | 12 | 1628 | |||
XM_005263769.3 | XP_005263826.1 | 12 | 1549 | |||
XM_005263770.3 | XP_005263827.1 | 12 | 1625 | |||
XM_006712748.3 | XP_006712811.1 | 12 | 1838 | |||
XM_006712749.3 | XP_006712812.1 | 12 | 1838 | |||
XM_017004903.1 | XP_016860392.1 | 12 | 1628 | |||
XM_017004904.1 | XP_016860393.1 | 12 | 1628 | |||
XM_017004905.1 | XP_016860394.1 | 12 | 1628 | |||
GenBank mRNA | AK022823.1 | - | 2 | 195 | ||
AK055979.1 | - | 12 | 1591 | |||
AK092234.1 | - | 6 | 739 | |||
AK315185.1 | - | 12 | 1678 | |||
AL136833.1 | - | 12 | 1676 | |||
AY220791.1 | - | 12 | 1546 | |||
BC017453.1 | - | 5 | 483 | |||
BC117255.1 | - | 12 | 1644 | 40125806 | ||
BC143898.1 | - | 12 | 1644 | 9052416 |