Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319987.1 | NP_001306916.1 | 2, 3 | 264 | ||
NM_024832.4 | NP_079108.3 | 3, 4 | 517 | |||
XM_011537164.2 | XP_011535466.1 | 2, 3 | 252 | |||
XM_017021651.1 | XP_016877140.1 | 3, 4 | 399 | |||
XM_017021652.1 | XP_016877141.1 | 3, 4 | 525 | |||
XM_017021653.1 | XP_016877142.1 | 3, 4 | 525 | |||
XM_017021654.1 | XP_016877143.1 | 3, 4 | 525 | |||
GenBank mRNA | AB081753.1 | - | 3, 4 | 365 | ||
AK021762.1 | - | 3, 4 | 517 | |||
AK128338.1 | - | 2, 3 | 264 | |||
BC070062.1 | - | 3, 4 | 505 | 30332851 | ||
BX248258.1 | - | 3, 4 | 503 | |||
DR003779.1 | - | 3, 4 | 557 |