Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031722.3 | NP_001026892.1 | 2 | 404 | ||
NM_001190724.2 | NP_001177653.1 | 2 | 404 | |||
NM_001254952.2 | NP_001241881.1 | 2 | 166 | |||
NM_001318762.1 | NP_001305691.1 | 2 | 166 | |||
NM_001318763.1 | NP_001305692.1 | 2 | 166 | |||
NM_024909.3 | NP_079185.2 | 2 | 166 | |||
NR_033823.2 | 2 | 166 | ||||
XM_005249415.2 | XP_005249472.1 | 2 | 367 | |||
XM_017011316.1 | XP_016866805.1 | 2 | 367 | |||
GenBank mRNA | AB075512.1 | - | 2 | 122 | ||
AK023220.1 | - | 2 | 132 | |||
AK225107.1 | - | 2 | 130 | |||
AK297661.1 | - | 2 | 287 | |||
AK298743.1 | - | 2 | 132 | |||
AL535697.3 | - | 2 | 140 | |||
AL833858.1 | - | 2 | 118 | |||
BC006105.1 | - | 2 | 177 | 2820493 | ||
BC025755.1 | - | 2 | 76 | |||
BC047303.1 | - | 2 | 404 | 4914615 | ||
BP202090.1 | - | 2 | 166 | |||
BX442806.2 | - | 2 | 113 | |||
CX786272.1 | - | 2 | 88 |