Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CDH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016279.3 | NP_057363.3 | 3 | 564 | ||
GenBank mRNA | AB035302.1 | - | 3 | 549 | ||
AK307779.1 | - | 3 | 564 | |||
AK308697.1 | - | 3 | 568 | |||
BC107594.1 | - | 3 | 526 | 40011942 | ||
BC113745.1 | - | 3 | 526 | 40011941 | ||
DA165253.1 | - | 3 | 564 |