Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 2 | 133 | ||
NM_001171944.1 | NP_001165415.1 | 3 | 192 | |||
NM_001171945.1 | NP_001165416.1 | 4 | 412 | |||
NM_001171946.1 | NP_001165417.1 | 3 | 180 | |||
NM_025154.5 | NP_079430.3 | 2 | 95 | |||
GenBank mRNA | AB018353.1 | - | 3 | 150 | ||
AB648918.1 | - | 2 | 113 | |||
AK022469.1 | - | 3 | 160 | |||
AK022816.1 | - | 2 | 87 | |||
AK302896.1 | - | 3 | 164 | |||
AK309120.1 | - | 4 | 384 | |||
BC013613.1 | - | 3 | 180 | |||
BC152418.1 | - | 2 | 150 | 8862528 | ||
BX538211.1 | - | 3 | 161 | |||
DA112466.1 | - | 3 | 192 | |||
DA666453.1 | - | 3 | 193 | |||
DA738650.1 | - | 2 | 95 |