Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308189.1 | NP_001295118.1 | 8 | 3156 | ||
NM_001308191.1 | NP_001295120.1 | 7 | 3132 | |||
NM_001308192.1 | NP_001295121.1 | 8 | 785 | |||
NM_001308193.1 | NP_001295122.1 | 7 | 734 | |||
NM_030627.3 | NP_085130.2 | 9 | 3207 | |||
XM_005265994.1 | XP_005266051.1 | 8 | 3183 | |||
GenBank mRNA | AB051460.1 | - | 8 | 1818 | ||
AK310905.1 | - | 7 | 1929 | |||
BC036899.1 | - | 7 | 718 | |||
BC117150.1 | - | 9 | 2195 | 40125701 | ||
BC143958.1 | - | 8 | 2144 | 9052476 | ||
BC143959.1 | - | 7 | 2120 | 9052477 | ||
BX401008.2 | - | 8 | 785 | |||
BX538213.1 | - | 9 | 2349 |