Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM25 SIRNA Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.11: 118531088 - 118546670 on Build GRCh38
UniGene ID:
Hs.564188
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241902
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144034.1
NP_001137506.1
5
869
NM_001144035.1
NP_001137507.1
5
866
NM_001144036.1
NP_001137508.1
4
557
NM_001144037.1
NP_001137509.1
6
1001
NM_001144038.1
NP_001137510.1
5
869
NM_001318755.1
NP_001305684.1
6
1001
NM_001318757.1
NP_001305686.1
5
710
NM_032780.3
NP_116169.2
6
1001
NR_134852.1
3
460
XM_005271703.1
XP_005271760.1
6
910
XM_006718926.2
XP_006718989.1
6
885
XM_006718927.2
XP_006718990.1
5
753
XM_011543037.2
XP_011541339.1
6
885
XM_011543038.2
XP_011541340.1
5
753
XM_017018422.1
XP_016873911.1
6
910
XM_017018423.1
XP_016873912.1
6
910
XM_017018424.1
XP_016873913.1
6
791
XM_017018425.1
XP_016873914.1
6
814
XM_017018426.1
XP_016873915.1
5
778
XM_017018427.1
XP_016873916.1
6
791
XM_017018428.1
XP_016873917.1
6
814
XM_017018429.1
XP_016873918.1
6
791
XM_017018430.1
XP_016873919.1
5
753
XM_017018431.1
XP_016873920.1
5
753
XM_017018432.1
XP_016873921.1
5
659
XM_017018433.1
XP_016873922.1
5
682
GenBank mRNA
AK027305.1
-
6
890
AK056325.1
-
4
443
AK096260.1
-
3
1573
AK124814.1
-
3
1719
AK225415.1
-
6
887
AK292359.1
-
5
753
AK293670.1
-
5
606
AY358919.1
-
5
754
BC042896.1
-
6
880
5168151
BC051841.1
-
4
737
5725777
DA760733.1
-
3
346
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0