Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 8 | 1352 | ||
XM_005251318.3 | XP_005251375.1 | 7 | 1141 | |||
XM_005251319.4 | XP_005251376.1 | 6 | 835 | |||
XM_005251320.3 | XP_005251377.1 | 5 | 627 | |||
XM_005251322.3 | XP_005251379.1 | 4 | 441 | |||
XM_017013920.1 | XP_016869409.1 | 9 | 1300 | |||
XM_017013921.1 | XP_016869410.1 | 9 | 1300 | |||
XM_017013922.1 | XP_016869411.1 | 9 | 1300 | |||
XM_017013923.1 | XP_016869412.1 | 8 | 1127 | |||
XM_017013924.1 | XP_016869413.1 | 9 | 1300 | |||
XM_017013925.1 | XP_016869414.1 | 6 | 846 | |||
XM_017013926.1 | XP_016869415.1 | 6 | 829 | |||
XM_017013927.1 | XP_016869416.1 | 7 | 857 | |||
XM_017013928.1 | XP_016869417.1 | 4 | 309 | |||
GenBank mRNA | AB051551.1 | - | 7 | 1973 | ||
AK056185.1 | - | 7 | 1315 | |||
AK300874.1 | - | 7 | 996 | |||
AK302648.1 | - | 8 | 1202 | |||
AK307350.1 | - | 4 | 416 | |||
AK307443.1 | - | 5 | 608 | |||
AK307636.1 | - | 7 | 986 | |||
BC030701.2 | - | 8 | 1225 | 4837403 |