Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152517.2 | NP_689730.2 | 1 | 1849 | ||
GenBank mRNA | AK055552.1 | - | 2 | 1748 | ||
AK296828.1 | - | 2 | 1684 | |||
BC033795.1 | - | 1 | 1833 | 5175299 | ||
BX647620.1 | - | 1 | 1382 |