Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178829.4 | NP_849151.2 | 1, 2 | 257 | ||
GenBank mRNA | AB083484.1 | - | 1, 2 | 205 | ||
AY927625.1 | - | 1 | 481 | |||
BC014596.1 | - | 1, 2 | 262 | 4825428 | ||
KU178863.1 | - | 1, 2 | 141 | |||
KU178864.1 | - | 1 | 728 |