Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194279.3 | NP_919255.2 | 3 | 298 | ||
GenBank mRNA | AK290728.1 | - | 3 | 271 | ||
BC015771.1 | - | 3 | 258 | 4849790 | ||
BC032893.2 | - | 3 | 253 | 5259721 | ||
BX248252.1 | - | 3 | 256 |