Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308195.1 | NP_001295124.1 | 10 | 2744 | ||
NM_001308196.1 | NP_001295125.1 | 12 | 2952 | |||
NM_001308200.1 | NP_001295129.1 | 7 | 1200 | |||
NM_198567.5 | NP_940969.3 | 9 | 1442 | |||
NR_131772.1 | 9 | 1442 | ||||
XM_011534553.2 | XP_011532855.1 | 11 | 6749 | |||
XM_011534554.2 | XP_011532856.1 | 11 | 2621 | |||
XM_011534556.2 | XP_011532858.1 | 10 | 6618 | |||
XM_017009454.1 | XP_016864943.1 | 10 | 2552 | |||
XM_017009455.1 | XP_016864944.1 | 11 | 2862 | |||
XM_017009456.1 | XP_016864945.1 | 11 | 2758 | |||
GenBank mRNA | BC032390.1 | - | 7 | 1168 | ||
BC037298.1 | - | 12 | 2953 | 4823991 | ||
BC066980.1 | - | 9 | 1435 | 4821958 | ||
BC073880.1 | - | 7 | 1200 | |||
KF151170.1 | - | 10 | 2612 |