Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 3 | 618 | ||
XM_006717317.3 | XP_006717380.1 | 3 | 644 | |||
XM_011519182.2 | XP_011517484.1 | 2 | 212 | |||
GenBank mRNA | AK095393.1 | - | 3 | 359 | ||
AK128529.1 | - | 1 | 453 | |||
AL832421.1 | - | 3 | 387 | |||
BC106035.1 | - | 3 | 330 | |||
BC136391.1 | - | 2 | 185 | |||
BX333979.2 | - | 3 | 617 |