Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 5 | 958 | ||
XM_006717317.3 | XP_006717380.1 | 5 | 984 | |||
XM_011519182.2 | XP_011517484.1 | 4 | 552 | |||
GenBank mRNA | AK074143.1 | - | 3 | 335 | ||
AK095393.1 | - | 5 | 699 | |||
AK128529.1 | - | 3 | 793 | |||
AL832421.1 | - | 5 | 727 | |||
BC106035.1 | - | 5 | 670 | |||
BC136391.1 | - | 4 | 525 |