Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYADML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003143.2 | 1 | 711 | |||
GenBank mRNA | AK057470.1 | - | 1 | 698 | ||
BC031088.1 | - | 1 | 701 | |||
BC038980.1 | - | 1 | 672 |