Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270398.1 | NP_001257327.1 | 6 | 1045 | ||
NM_002821.4 | NP_002812.2 | 6 | 1172 | |||
NM_152880.3 | NP_690619.1 | 6 | 1172 | |||
NM_152881.3 | NP_690620.1 | 6 | 1172 | |||
NM_152882.3 | NP_690621.1 | 6 | 1172 | |||
NR_072997.1 | 5 | 981 | ||||
NR_072998.1 | 6 | 1172 | ||||
XM_011514765.2 | XP_011513067.1 | 6 | 1040 | |||
XM_011514766.2 | XP_011513068.1 | 6 | 1040 | |||
GenBank mRNA | AB209350.1 | - | 6 | 1071 | ||
AF531868.1 | - | 6 | 1098 | |||
AF531869.1 | - | 6 | 1098 | |||
AF531870.1 | - | 6 | 1098 | |||
AF531871.1 | - | 6 | 1098 | |||
AF531872.1 | - | 6 | 1098 | |||
AK291016.1 | - | 6 | 1004 | |||
AK292589.1 | - | 6 | 1099 | |||
AK296953.1 | - | 6 | 980 | |||
BC046109.2 | - | 6 | 1112 | |||
BC071557.1 | - | 6 | 1126 | 30347652 | ||
U33635.1 | - | 6 | 1143 | |||
U40271.2 | - | 6 | 1098 |