Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000152.4 | NP_000143.2 | 2, 3 | 945 | ||
NM_001079803.2 | NP_001073271.1 | 3, 4 | 760 | |||
NM_001079804.2 | NP_001073272.1 | 2, 3 | 680 | |||
XM_005257193.2 | XP_005257250.1 | 3, 4 | 746 | |||
XM_005257194.4 | XP_005257251.1 | 3, 4 | 786 | |||
GenBank mRNA | AK299462.1 | - | 724 | |||
BC040431.1 | - | 3, 4 | 692 | 5739991 | ||
BF206270.1 | - | 1, 2 | 467 | |||
DQ907243.1 | - | 1, 2 | 548 | |||
M34424.1 | - | 1, 2 | 580 | |||
Y00839.1 | - | 2, 3 | 767 |