Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001084392.1 | NP_001077861.1 | 3 | 423 | ||
NM_001355.3 | NP_001346.1 | 4 | 474 | |||
GenBank mRNA | AK292044.1 | - | 3 | 367 | ||
BC005971.1 | - | 3 | 371 | 4082777 | ||
BC015508.2 | - | 3 | 392 | 3902943 | ||
CR456431.1 | - | 3 | 369 | |||
U49785.1 | - | 4 | 472 | |||
U84143.1 | - | 3 | 360 |