Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF089106.1 | - | 628 | |||
AF161524.1 | - | 587 | ||||
AX771012.1 | - | 628 | ||||
AX771013.1 | - | 587 |