Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002929.2 | NP_002920.1 | 6 | 1484 | ||
XM_017020684.1 | XP_016876173.1 | 6 | 3151 | |||
GenBank mRNA | AK289912.1 | - | 6 | 1608 | ||
BC140945.1 | - | 6 | 1449 | 9056757 | ||
BM702747.1 | - | 1 | 73 | |||
L77503.1 | - | 6 | 1391 | |||
U63973.1 | - | 6 | 1484 |