Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320800.1 | NP_001307729.1 | 2 | 378 | ||
NM_007170.2 | NP_009101.2 | 3 | 686 | |||
XM_005270355.3 | XP_005270412.1 | 3 | 658 | |||
XM_006710287.2 | XP_006710350.1 | 4 | 726 | |||
XM_011540497.1 | XP_011538799.1 | 5 | 759 | |||
XM_011540500.2 | XP_011538802.1 | 3 | 658 | |||
XM_017000047.1 | XP_016855536.1 | 4 | 891 | |||
XM_017000048.1 | XP_016855537.1 | 3 | 860 | |||
XM_017000049.1 | XP_016855538.1 | 3 | 860 | |||
GenBank mRNA | AB057597.1 | - | 3 | 678 | ||
AJ132545.1 | - | 3 | 678 | |||
AK027573.1 | - | 3 | 660 | |||
AK294172.1 | - | 2 | 353 | |||
AK299269.1 | - | 3 | 681 | |||
AK302494.1 | - | 3 | 718 | |||
BC022194.1 | - | 3 | 666 | |||
BC033085.1 | - | 2 | 297 | 5425494 |