Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK2 SIRNA Assays ›
Gene Symbol:
MARK2
Gene Name:
microtubule affinity regulating kinase 2
Gene Aliases:
EMK-1, EMK1, PAR-1, Par-1b, Par1b
Chromosome Location:
Chr.11: 63838928 - 63911020 on Build GRCh38
UniGene ID:
Hs.567261
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
242440
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039469.2
NP_001034558.2
6, 7
1047
NM_001163296.1
NP_001156768.1
6, 7
1047
NM_001163297.1
NP_001156769.1
6, 7
1047
NM_004954.4
NP_004945.4
6, 7
1047
NM_017490.3
NP_059672.2
6, 7
864
XM_011544789.1
XP_011543091.1
7, 8
938
XM_011544790.1
XP_011543092.1
7, 8
938
XM_011544791.1
XP_011543093.1
7, 8
938
XM_011544792.1
XP_011543094.1
7, 8
938
XM_011544793.1
XP_011543095.1
7, 8
938
XM_011544794.1
XP_011543096.1
7, 8
938
XM_011544795.1
XP_011543097.1
7, 8
938
XM_011544796.1
XP_011543098.1
7, 8
938
XM_011544797.1
XP_011543099.1
7, 8
938
XM_011544798.1
XP_011543100.1
7, 8
938
XM_017017310.1
XP_016872799.1
6, 7
1020
XM_017017311.1
XP_016872800.1
7, 8
938
GenBank mRNA
AB188493.1
-
6, 7
1051
AB271694.1
-
5, 6
369
AF387638.1
-
5, 6
387
AK225766.1
-
6, 7
882
AK290339.1
-
6, 7
832
AK307858.1
-
7, 8
683
BC008771.2
-
6, 7
439
BC084540.1
-
6, 7
506
4591688
BT007342.1
-
5, 6
369
X97630.1
-
6, 7
776
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
mitophagy
neuron migration
protein phosphorylation
positive regulation of neuron projection development
Wnt signaling pathway
peptidyl-threonine phosphorylation
establishment of cell polarity
activation of protein kinase activity
intracellular signal transduction
establishment or maintenance of epithelial cell apical/basal polarity
protein autophosphorylation
regulation of axonogenesis
regulation of cytoskeleton organization
mitochondrion localization
cell-cell adhesion
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
lipid binding
protein kinase activator activity
poly(A) RNA binding
tau-protein kinase activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline