Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 9 | 1157 | ||
NM_018401.2 | NP_060871.1 | 8 | 1047 | |||
XM_005247982.3 | XP_005248039.1 | 3 | 1554 | |||
XM_005247983.4 | XP_005248040.1 | 3 | 812 | |||
XM_011513497.1 | XP_011511799.1 | 7 | 1151 | |||
XM_011513501.2 | XP_011511803.1 | 4 | 346 | |||
XM_011513504.1 | XP_011511806.1 | 3 | 200 | |||
XM_017008374.1 | XP_016863863.1 | 7 | 648 | |||
XM_017008375.1 | XP_016863864.1 | 8 | 844 | |||
XM_017008377.1 | XP_016863866.1 | 3 | 1599 | |||
GenBank mRNA | AJ250839.1 | - | 8 | 764 | ||
AK292696.1 | - | 8 | 1014 | |||
AK313846.1 | - | 8 | 1049 | |||
AL832436.1 | - | 2 | 100 | |||
AY358353.1 | - | 8 | 791 | |||
BC038238.1 | - | 8 | 741 | 5744969 | ||
BC069203.1 | - | 9 | 1078 |