Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 13 | 3054 | ||
NM_018401.2 | NP_060871.1 | 12 | 2944 | |||
XM_005247982.3 | XP_005248039.1 | 7 | 3451 | |||
XM_005247983.4 | XP_005248040.1 | 7 | 2709 | |||
XM_011513497.1 | XP_011511799.1 | 11 | 3048 | |||
XM_011513501.2 | XP_011511803.1 | 8 | 2243 | |||
XM_011513504.1 | XP_011511806.1 | 7 | 2097 | |||
XM_017008377.1 | XP_016863866.1 | 7 | 3496 | |||
GenBank mRNA | AJ250839.1 | - | 12 | 2661 | ||
AK055597.1 | - | 3 | 1591 | |||
AL832436.1 | - | 6 | 2000 | |||
AY358353.1 | - | 12 | 2691 | |||
BC020281.1 | - | 1 | 1006 | |||
BC024977.1 | - | 1 | 1006 | |||
BC038238.1 | - | 12 | 2638 | 5744969 |